(S)-3-(Boc-amino)-3-(4-cyanophenyl)propionic acid 98%

Supplier: Ambeed
Warning

A395100-100MG A395100-1G A395100-250MG A395100-50MG
76990-178EA 11 USD
76990-178 76990-180 76990-182 76990-184
(S)-3-(Boc-amino)-3-(4-cyanophenyl)propionic acid 98%
(S)-3-(Boc-amino)-3-(4-cyanophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₄
MDL Number: MFCD03427930
CAS Number: 500770-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR