Ethyl 3-bromo-4-iodobenzoate 97%

Supplier: Ambeed
Warning

A394611-1G A394611-250MG
77016-238EA 37 USD
77016-238 77016-240
Ethyl 3-bromo-4-iodobenzoate 97%
Ethyl 3-bromo-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrIO₂
MDL Number: MFCD11520423
CAS Number: 386267-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR