Ethyl-3-cyanopropanoate 97%

Supplier: Ambeed
Warning

A394607-1G A394607-250MG A394607-5G A394607-100MG
77016-232EA 62 USD
77016-232 77016-234 77016-236 77890-813
Ethyl-3-cyanopropanoate 97%
Ethyl-3-cyanopropanoate

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MDL Number: MFCD01695650
CAS Number: 10137-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR