(-)-Arctiin 98%

Supplier: Ambeed
Warning

A394382-10MG A394382-1MG A394382-50MG A394382-100MG A394382-1G A394382-250MG
77003-790EA 8 USD
77003-790 77003-792 77003-794 77601-734 77882-053 77882-054
(-)-Arctiin 98%
(-)-Arctiin

Precaución: for Research Use Only

Formula: C₂₇H₃₄O₁₁
MDL Number: MFCD00870630
CAS Number: 20362-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR