tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

A394315-100G A394315-10G A394315-1G A394315-250MG A394315-25G A394315-5G
77003-778EA 290.69 USD
77003-778 77003-780 77003-782 77003-784 77003-786 77003-788
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.3 g/mol
MDL Number: MFCD22409620
CAS Number: 143557-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR