(1H-Benzo[d][1,2,3]triazol-1-yl)(phenyl)methanone 95%

Supplier: Ambeed
Warning

A393495-1G A393495-250MG
76990-174EA 29.71 USD
76990-174 76990-176
(1H-Benzo[d][1,2,3]triazol-1-yl)(phenyl)methanone 95%
(1H-Benzo[d][1,2,3]triazol-1-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₉N₃O
MW: 223.23 g/mol
MDL Number: MFCD00451114
CAS Number: 4231-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR