4,7-Dibromo-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A393359-5G A393359-1G
76990-172EA 816.9 USD
76990-172 77684-126
4,7-Dibromo-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole 95%, Ambeed.Inc
4,7-Dibromo-2-(heptadecan-9-yl)-2H-benzo[d][1,2,3]triazole

Precaución: For research use only.

Formula: C₂₃H₃₇Br₂N₃
MW: 515.36 g/mol
CAS Number: 1312118-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR