1,2,3,4-Tetrahydroquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A393112-1G A393112-250MG
77016-212EA 152.77 USD
77016-212 77684-123
1,2,3,4-Tetrahydroquinoline-3-carboxylic acid 97%
1,2,3,4-Tetrahydroquinoline-3-carboxylic acid

Precaución: For research use only.

Formula: C₁₀H₁₁NO₂
MW: 177.19 g/mol
MDL Number: MFCD06739005
CAS Number: 114527-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR