4-Bromo-2,6-diiodophenol 97%

Supplier: Ambeed
Warning

A393105-100MG A393105-10G A393105-1G A393105-250MG
76990-164EA 9 USD
76990-164 76990-166 76990-168 76990-170
4-Bromo-2,6-diiodophenol 97%
4-Bromo-2,6-diiodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrI₂O
MDL Number: MFCD00672916
CAS Number: 15459-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR