Vatalanib dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Chlorophenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine dihydrochloride, Vatalanib 2HCl

A392716-100MG A392716-1MG A392716-50MG A392716-10MG A392716-250MG
77033-568EA 115 USD
77033-568 77033-570 77033-572 77601-728 77872-611
Vatalanib dihydrochloride 98%
Vatalanib dihydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₁₇Cl₃N₄
MW: 419.7348 g/mol
MDL Number: MFCD08458964
CAS Number: 212141-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR