2-(Benzyloxy)-4-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A392294-100MG A392294-250MG
76990-152EA 16 USD
76990-152 76990-154
2-(Benzyloxy)-4-fluorobenzaldehyde 95%
2-(Benzyloxy)-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MDL Number: MFCD12025063
CAS Number: 202857-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR