Ethyl 4-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A392118-100G A392118-1G A392118-25G A392118-5G
77016-202EA 369.2 USD
77016-202 77016-204 77016-206 77016-208
Ethyl 4-bromo-2-methylbenzoate 98%
Ethyl 4-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.09 g/mol
MDL Number: MFCD11042853
CAS Number: 220389-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR