3-Fluoropiperidine hydrochloride 98%

Supplier: Ambeed
Warning

A391886-1G A391886-250MG A391886-5G
77016-196EA 56 USD
77016-196 77016-198 77016-200
3-Fluoropiperidine hydrochloride 98%
3-Fluoropiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀FN·HCl
MDL Number: MFCD03452799
CAS Number: 737000-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR