2-Fluoro-5-hydroxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A391594-100MG
A391594-1G
A391594-250MG
A391594-25G
A391594-5G
77033-548EA
7
USD
77033-548
77033-550
77033-552
77033-554
77033-556
2-Fluoro-5-hydroxybenzaldehyde 98%
2-Fluoro-5-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅FO₂ |
MDL Number:
MFCD09038469 CAS Number: 103438-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...