6-Fluoro-8-nitroquinoline 95+%

Supplier: Ambeed
Warning

A391276-100MG A391276-1G A391276-250MG
77016-184EA 23.34 USD
77016-184 77016-186 77016-188
6-Fluoro-8-nitroquinoline 95+%
6-Fluoro-8-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅FN₂O₂
MDL Number: MFCD09907841
CAS Number: 343-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR