5-Hydroxy-2-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A391237-100MG A391237-1G
76990-134EA 75.33 USD
76990-134 76990-136
5-Hydroxy-2-methoxybenzoic acid 95%
5-Hydroxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.14 g/mol
MDL Number: MFCD10698756
CAS Number: 61227-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR