2-Chloro-5-nitrothiazole 97%

Supplier: Ambeed
Warning

A391086-10G A391086-1G A391086-5G A391086-250MG
77003-752EA 214 USD
77003-752 77003-754 77003-756 77882-044
2-Chloro-5-nitrothiazole 97%
2-Chloro-5-nitrothiazole

Precaución: for Research Use Only

Formula: C₃HClN₂O₂S
MDL Number: MFCD09743979
CAS Number: 3034-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR