2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,4,4-Tetramethylcyclobutane-1,3-diol

A390882-25G A390882-5G
77016-178EA 61 USD
77016-178 77016-180
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) 98%
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers)

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00001329
CAS Number: 3010-96-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR