(4-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A390782-1G A390782-250MG A390782-5G
77016-172EA 34 USD
77016-172 77016-174 77016-176
(4-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid 96%
(4-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₃
MDL Number: MFCD08276834
CAS Number: 1072951-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR