5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A389928-100MG
A389928-1G
A389928-250MG
76990-128EA
80.63
USD
76990-128
76990-130
76990-132
5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇ClN₂ MW: 166.60 g/mol |
MDL Number:
MFCD11040729 CAS Number: 145934-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...