3-(1H-Pyrazol-3-yl)aniline 96%

Supplier: Ambeed
Danger

A389658-100MG A389658-1G A389658-1MG A389658-250MG
76990-118EA 55 USD
76990-118 76990-120 76990-122 76990-124
3-(1H-Pyrazol-3-yl)aniline 96%
3-(1H-Pyrazol-3-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD00665959
CAS Number: 89260-46-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR