3-Nitro-1H-pyrrolo[2,3-c]pyridine 95%

Supplier: Ambeed
Warning

A389415-100MG A389415-1G A389415-250MG A389415-5G
76990-110EA 32.89 USD
76990-110 76990-112 76990-114 76990-116
3-Nitro-1H-pyrrolo[2,3-c]pyridine 95%
3-Nitro-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.13 g/mol
MDL Number: MFCD11846274
CAS Number: 67058-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR