Ethyl 6-chloro-2-methylpyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A389200-100MG A389200-250MG A389200-5G A389200-1G
76990-098EA 18 USD
76990-098 76990-100 77613-176 77890-804
Ethyl 6-chloro-2-methylpyrimidine-4-carboxylate 98%
Ethyl 6-chloro-2-methylpyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD16987970
CAS Number: 744253-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR