6-Methoxyquinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-6-methoxyquinoline

A389135-100MG A389135-1G A389135-250MG A389135-5G
77016-156EA 7.43 USD
77016-156 77016-158 77016-160 77016-162
6-Methoxyquinoline-3-carboxylic acid 95%
6-Methoxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.194 g/mol
MDL Number: MFCD08437579
CAS Number: 71082-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR