4-(4-Morpholinyl)-1-butanol 95%

Supplier: Ambeed
Warning

A388818-250MG A388818-5G A388818-100MG
77003-740EA 58 USD
77003-740 77672-934 77882-040
4-(4-Morpholinyl)-1-butanol 95%
4-(4-Morpholinyl)-1-butanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂
MDL Number: MFCD03211280
CAS Number: 5835-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR