2,3,5-Trichloro-4-formylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-Trichloroisonicotinaldehyde

A388569-1G A388569-250MG
77003-730EA 11.67 USD
77003-730 77003-732
2,3,5-Trichloro-4-formylpyridine 95%
2,3,5-Trichloro-4-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO
MW: 210.4500 g/mol
MDL Number: MFCD06410668
CAS Number: 251997-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR