2-Methyl-1,3,4-oxadiazole 95%

Supplier: Ambeed
Warning

A388559-100MG A388559-1G A388559-250MG A388559-5G
77033-524EA 37.14 USD
77033-524 77033-526 77033-528 77033-530
2-Methyl-1,3,4-oxadiazole 95%
2-Methyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₃H₄N₂O
MW: 84.07 g/mol
MDL Number: MFCD16877047
CAS Number: 3451-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR