Methyl 3-bromo-5-fluoro-4-nitrobenzoate 95%

Supplier: Ambeed
Warning

A388522-100MG A388522-10G A388522-1G A388522-250MG A388522-25G A388522-5G
77016-144EA 50 USD
77016-144 77016-146 77016-148 77016-150 77016-152 77016-154
Methyl 3-bromo-5-fluoro-4-nitrobenzoate 95%
Methyl 3-bromo-5-fluoro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₄
MDL Number: MFCD14635859
CAS Number: 1123171-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR