2-(2-Chlorophenoxy)-4-methylaniline 97%

Supplier: Ambeed
Warning

A388293-1G A388293-250MG
77003-728EA 338 USD
77003-728 77882-037
2-(2-Chlorophenoxy)-4-methylaniline 97%
2-(2-Chlorophenoxy)-4-methylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD08687430
CAS Number: 946728-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR