2-Methoxy-4-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A388273-1G A388273-250MG A388273-5G
76990-086EA 60.48 USD
76990-086 76990-088 77684-119
2-Methoxy-4-(trifluoromethyl)benzoic acid 98%
2-Methoxy-4-(trifluoromethyl)benzoic acid

Precaución: For research use only.

Formula: C₉H₇F₃O₃
MDL Number: MFCD04115976
CAS Number: 448-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR