4-(tert-Butoxymethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(tert-Butoxymethyl)phenyl)boronic acid

A387811-1G A387811-250MG A387811-5G
77003-718EA 94.43 USD
77003-718 77003-720 77003-722
4-(tert-Butoxymethyl)phenylboronic acid 98%
4-(tert-Butoxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
MW: 208.07 g/mol
MDL Number: MFCD03701685
CAS Number: 1024017-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR