2-Aminomethyl-1H-imidazole dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminomethyl-1H-imidazole dihydrochloride, 2-(Aminomethyl)imidazole dihydrochloride, (1H-Imidazol-2-yl)methanamine dihydrochloride
A387408-1G
A387408-250MG
A387408-5G
77016-134EA
63
USD
77016-134
77016-136
77892-794
2-Aminomethyl-1H-imidazole dihydrochloride 98%
2-Aminomethyl-1H-imidazole dihydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉Cl₂N₃ |
MDL Number:
MFCD06738779 CAS Number: 22600-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...