4-Hydroxy-2-(methylthio)pyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A387304-10G A387304-1G A387304-250MG A387304-5G
77003-710EA 129.43 USD
77003-710 77003-712 77003-714 77003-716
4-Hydroxy-2-(methylthio)pyrimidine-5-carbonitrile 97%
4-Hydroxy-2-(methylthio)pyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃OS
MW: 167.18 g/mol
MDL Number: MFCD00194951
CAS Number: 89487-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR