Pixantrone maleate 98%

Supplier: Ambeed
Warning

Synonyms: Pixantrone, 6,9-Bis((2-aminoethyl)amino)benzo[g]isoquinoline-5,10-dione dimaleate

A387062-10MG A387062-1MG A387062-50MG A387062-5MG
77033-506EA 98 USD
77033-506 77033-508 77033-510 77033-512
Pixantrone maleate 98%
Pixantrone maleate

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₅O₂·C₈H₈O₈
MW: 557.52 g/mol
MDL Number: MFCD00920108
CAS Number: 144675-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR