2,2,2-Trifluoro-1-pyridin-3-ylethylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-1-(pyridin-3-yl)ethanamine dihydrochloride

A387024-100MG A387024-250MG
77033-502EA 61.54 USD
77033-502 77033-504
2,2,2-Trifluoro-1-pyridin-3-ylethylamine hydrochloride 97%
2,2,2-Trifluoro-1-pyridin-3-ylethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂F₃N₂
MW: 249.06 g/mol
MDL Number: MFCD06738892
CAS Number: 1159825-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR