2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile 98%

Supplier: Ambeed
Warning

A386971-100MG A386971-1G A386971-250MG
76990-068EA 31.83 USD
76990-068 76990-070 76990-072
2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile 98%
2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO
MW: 307.42 g/mol
MDL Number: MFCD26393887
CAS Number: 1246213-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR