6-Bromo-o-toluonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methylbenzonitrile

A386898-100G A386898-100MG A386898-10G A386898-1G A386898-250MG A386898-25G A386898-5G
77033-488EA 988 USD
77033-488 77033-490 77033-492 77033-494 77033-496 77033-498 77033-500
6-Bromo-o-toluonitrile 98%
6-Bromo-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.0439 g/mol
MDL Number: MFCD09834775
CAS Number: 77532-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR