[4-(1-Methoxyethyl)phenyl]boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: (4-(1-Methoxyethyl)phenyl)boronic acid

A386653-100MG A386653-1G A386653-250MG
77016-116EA 30 USD
77016-116 77016-118 77016-120
[4-(1-Methoxyethyl)phenyl]boronic acid 95+%
[4-(1-Methoxyethyl)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MDL Number: MFCD09864274
CAS Number: 945723-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR