6-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one 95%
Supplier: Ambeed
|
Warning
|
A386471-100MG
A386471-1G
A386471-250MG
77016-110EA
96
USD
77016-110
77016-112
77016-114
6-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one 95%
6-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂O |
MDL Number:
MFCD12024514 CAS Number: 676515-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...