Diethyl-3,3-diethoxypropane-1,1-dicarboxylate 97% (by GC)

Supplier: Ambeed
Warning

Synonyms: 3,3-Diethoxypropane-1,1-dicarboxylic Acid Diethyl Ester, Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate

A385308-1G A385308-250MG A385308-5G
77003-686EA 24.41 USD
77003-686 77003-688 77003-690
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate 97% (by GC)
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₆
MW: 276.33 g/mol
MDL Number: MFCD00671525
CAS Number: 21339-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR