(3-((4-(tert-Butyl)-2-methylphenoxy)methyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A385280-1G
A385280-250MG
A385280-5G
76990-046EA
37
USD
76990-046
76990-048
76990-050
(3-((4-(tert-Butyl)-2-methylphenoxy)methyl)phenyl)boronic acid 98%
(3-((4-(tert-Butyl)-2-methylphenoxy)methyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₈H₂₃BO₃ |
MDL Number:
MFCD09038420 CAS Number: 1072951-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...