N-(Hydroxymethyl)phthalimide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Hydroxymethyl)isoindoline-1,3-dione
A384940-1000G
A384940-100G
A384940-10G
A384940-25G
A384940-500G
77003-676EA
47
USD
77003-676
77003-678
77003-680
77003-682
77003-684
N-(Hydroxymethyl)phthalimide 97%
N-(Hydroxymethyl)phthalimide
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ |
MDL Number:
MFCD00005899 CAS Number: 118-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...