4-Hydroxy-o-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid

A384671-100G A384671-10G A384671-1G A384671-25G A384671-5G
77033-456EA 199 USD
77033-456 77033-458 77033-460 77033-462 77033-464
4-Hydroxy-o-toluic acid 98%
4-Hydroxy-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD02182261
CAS Number: 578-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR