4-Morpholinobenzaldehyde 97%

Supplier: Ambeed
Warning

A384262-100G A384262-10G A384262-1G A384262-25G A384262-5G
77016-096EA 333.13 USD
77016-096 77016-098 77016-100 77016-102 77016-104
4-Morpholinobenzaldehyde 97%
4-Morpholinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.2264 g/mol
MDL Number: MFCD00735826
CAS Number: 1204-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR