3-Chloro-7-methoxy-1H-indazole 95+%

Supplier: Ambeed
Danger

A384219-100MG A384219-1G A384219-250MG
77016-090EA 10 USD
77016-090 77016-092 77016-094
3-Chloro-7-methoxy-1H-indazole 95+%
3-Chloro-7-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O
MDL Number: MFCD11846519
CAS Number: 1260758-52-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR