(5-Fluoro-2-methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Fluoro-2-methoxyphenyl)acetic acid

A384111-1G A384111-5G A384111-10G
77033-444EA 50.93 USD
77033-444 77033-446 77672-902
(5-Fluoro-2-methoxyphenyl)acetic acid 97%
(5-Fluoro-2-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17 g/mol
MDL Number: MFCD00671771
CAS Number: 383134-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR