2-Amino-9-((1S,3R,4S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-1H-purin-6(9H)-one 97%

Supplier: Ambeed
Warning

A383747-100MG A383747-1G A383747-1MG A383747-250MG A383747-5G
77033-442EA 25 USD
77033-442 77668-782 77668-784 77668-786 77872-603
2-Amino-9-((1S,3R,4S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-1H-purin-6(9H)-one 97%
2-Amino-9-((1S,3R,4S)-4-(benzyloxy)-3-((benzyloxy)methyl)-2-methylenecyclopentyl)-1H-purin-6(9H)-one

Precaución: for Research Use Only

Formula: C₂₆H₂₇N₅O₃
MDL Number: MFCD11977291
CAS Number: 142217-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR