5-Chloro-8-iodo-1,6-naphthyridine 95%

Supplier: Ambeed
Warning

A383352-100MG A383352-250MG
76990-034EA 88.06 USD
76990-034 76990-036
5-Chloro-8-iodo-1,6-naphthyridine 95%
5-Chloro-8-iodo-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄ClIN₂
MW: 290.49 g/mol
MDL Number: MFCD16250553
CAS Number: 107484-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR