7-Amino-2-benzoxazolinone 95%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-2-hydroxybenzoxazole, 7-Amino-2-benzoxazolone, 7-Aminobenzo[d]oxazol-2(3H)-one

A383012-100MG A383012-1G
76990-030EA 18.04 USD
76990-030 76990-032
7-Amino-2-benzoxazolinone 95%
7-Amino-2-benzoxazolinone

Precaución: for Research Use Only

Formula: C₇H₆N₂O₂
MW: 150.14 g/mol
MDL Number: MFCD09753616
CAS Number: 81282-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR