Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A382846-100MG A382846-10G A382846-1G A382846-250MG A382846-25G
76990-020EA 11.67 USD
76990-020 76990-022 76990-024 76990-026 76990-028
Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate 95%
Ethyl 6-(4-aminophenyl)-1-(4-methoxyphenyl)-7-oxo-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₄O₄
MW: 406.43 g/mol
MDL Number: MFCD18251628
CAS Number: 503615-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR